
4-Picoline N-oxide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1003-67-4
:
Formula:C₆H₇NO
MDL Number:MFCD00006210
:109.13 g/mol
Boiling Pt:152 °C (11 mmHg)
Melting Pt:182…185 °C
Density:1,01…1.02 g/cm³ (185 °C)
Flash Pt:149 °C (300 °F)
EINECS:213-712-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together