4-Nitropyridine-N-oxide
Glass bottle for solids, 100 g
Catalog # MOLE14398574-100G
Marketsource
Supplier: Molekula
4-Nitropyridine-N-oxide
Catalog # MOLE14398574-100G
Supplier: Molekula
Marketsource
CAS Number: 1124-33-0
Jump to:
Product Details & Documents
Synonyms: 4-Nitropyridine N-oxide
- CAS Number:1124-33-0
- Formula:C₅H₄N₂O₃
- MDL Number:MFCD00006206
- Molecular Weight:140.1 g/mol
- Boiling Pt:397 °C (1013 hPa)
- Melting Pt:161 °C
- Density:1,52 g/cm³ (20 °C)
- EINECS:214-395-4
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



