Order Entry
Northern Ireland
ContactUsLinkComponent
 
Pyrazinamide for synthesis, Sigma-Aldrich®
  8.21050.0025
 :  Merck
 :  
Pyrazinamide for synthesis, Sigma-Aldrich®
  8.21050.0025
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Hill Formula:
    C₅H₅N₃O
  • Molar Mass:
    123.12 g/mol
  • Melting Point:
    188 - 191 °C
  • pH value:
    7 (H₂O)
  • Solubility:
    (20 °C) slightly soluble
  • Storage class:
    10 - 13 Other liquids and solids
  • WGK:
    WGK 2 water endangering

 

 

 :
 :123.11 g/mol
 :12,08140
 :357 °C (1013 hPa)
 :188…191 °C
 :M-Clarity™ quality level = MQ200
 :202-717-6
 :98-96-4
 :C₅H₅N₃O
 :MFCD00006132