

1,4-Butanesultone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1633-83-6
:
Formula:C₄H₈O₃S
MDL Number:MFCD00006584
:136.17 g/mol
Boiling Pt:153 °C (1013 hPa)
Melting Pt:12,5…15 °C
Density:1,331 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:216-647-9
:GHS08
:GHS07
Specification Test Results
- Solubility:54 g/l (20 °C) (decomposition)
- Melting Point:12.5 - 15.0 °C
- Molar Mass:136.16 g/mol
- Boiling Point:153 °C (19 hPa)
- Flash Point:154 °C
- Refractive Index:1.462 (25 °C, 589 nm)
- Density:1.34 g/cm3 (20 °C)
- Synonyms:4-Hydroxy-1-butanesulfonic acid δ-sultone
- Hill Formula:C4H8O3S
Specification Test Results
Frequently Bought Together