Order Entry
Northern Ireland
ContactUsLinkComponent
Cobalt (III) acetylacetonate for synthesis, Sigma-Aldrich®
 
Cobalt (III) acetylacetonate for synthesis, Sigma-Aldrich®
  8.02528.0050
 :  Merck
 :  
 
Cobalt (III) acetylacetonate for synthesis, Sigma-Aldrich®
  8.02528.0050
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

 

 

  • Solubility:
    3 g/l (30 °C)
  • Melting Point:
    196 - 202 °C
  • Molar Mass:
    356.26 g/mol
  • Bulk Density:
    830 kg/m3

 

 

 :
 :356.26 g/mol
 :M-Clarity™ quality level = MQ200
 :244-527-6
 :GHS08
 :Ambient
 :21679-46-9
 :C₁₅H₂₁CoO₆
 :MFCD00013488