Order Entry
Northern Ireland
ContactUsLinkComponent
Cobalt(II) acetylacetonate 99%
Cobalt(II) acetylacetonate 99%
Certificates
undefined
Cobalt(II) acetylacetonate 99%

Specification Test Results

  • Appearance:
    Purple Crystalline powder
  • Infrared spectrum:
    Conforms
  • Assay:
    98.5 to 101.5 % (ex Co)

Specification Test Results

Related Information

Frequently Bought Together