

Titanium(IV) ethanolate (≥33% TiO₂)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3087-36-3
:
Formula:C₈H₂₀O₄Ti
MDL Number:MFCD00009071
:228.12 g/mol
Boiling Pt:150…152 °C (10 mmHg)
Melting Pt:–40 °C
Density:1,088
Flash Pt:28 °C (84 °F)
Storage Temperature:Ambient
EINECS:221-410-8
UN:1993
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together