Order Entry
Northern Ireland
ContactUsLinkComponent
2-Hydroxy-1-naphthaldehyde
2-Hydroxy-1-naphthaldehyde
Catalog # MOLEM78626682
Supplier:  Molekula
CAS Number:  
2-Hydroxy-1-naphthaldehyde
Catalog # MOLEM78626682
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :172.18 g/mol
EINECS:211-902-0
CAS Number:708-06-5
Formula:C₁₁H₈O₂
MDL Number:MFCD00004005