Dicerium trisulphide ≥99.9% (metals basis)
: Cerium (III) sulphide, Cerous sulphide
:12014-93-6
:
:Ce₂S₃
:MFCD00016007
:5.02
:376.43
:2100 °C
:Ambient
:234-603-7
:3134
:4.3 III
:GHS02
:GHS07
- Environmentally Preferable:
Frequently Bought Together