Tetrabutylammonium hexafluorophosphate
Catalog # MOLE32102353-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:387.43 g/mol
EINECS:221-472-6
UN:3263
ADR:8,III
CAS Number:3109-63-5
Formula:C₁₆H₃₆F₆NP
MDL Number:MFCD00011748