Order Entry
Northern Ireland
ContactUsLinkComponent
Tetrabutylammonium hexafluorophosphate 98%
Irritant
Tetrabutylammonium hexafluorophosphate 98%
Certificates
undefined
Irritant
Tetrabutylammonium hexafluorophosphate 98%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    242 to 247 °C
  • Elemental Analysis:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together