Order Entry
Northern Ireland
ContactUsLinkComponent
Tetrapentylammonium iodide for synthesis, Sigma-Aldrich®
Irritant
Tetrapentylammonium iodide for synthesis, Sigma-Aldrich®
Catalog # 8.41762.0025
Supplier:  Merck
CAS Number:  
Irritant
Tetrapentylammonium iodide for synthesis, Sigma-Aldrich®
Catalog # 8.41762.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    7.40 g/l (25 °C)
  • Melting Point:
    135 - 137 °C
  • Molar Mass:
    425.47 g/mol

Specification Test Results

About this item

 :
 :425.48 g/mol
Melting Pt:135…137 °C
Density:1,18 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ200
EINECS:219-688-0
 :GHS07
CAS Number:2498-20-6
Formula:C₂₀H₄₄IN
MDL Number:MFCD00041980