Order Entry
Northern Ireland
ContactUsLinkComponent
 
Irritant
Tetraheptylammonium iodide for synthesis, Sigma-Aldrich®
Catalog # 8.41874.0005
Supplier:  Merck
CAS Number:  
Irritant
Tetraheptylammonium iodide for synthesis, Sigma-Aldrich®
Catalog # 8.41874.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Melting Point:
    121 - 124 °C
  • Molar Mass:
    537.69 g/mol

Specification Test Results

About this item

 :
 :537.7 g/mol
Melting Pt:124 °C
Density:1,066 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ200
EINECS:222-570-1
 :GHS07
CAS Number:3535-83-9
Formula:C₂₈H₆₀IN
MDL Number:MFCD00041984