

Antimony (III) sulphide ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1345-04-6
:
Formula:S₃Sb₂
MDL Number:MFCD00011217
:339.72 g/mol
Merck Index:14,713
Boiling Pt:1150 °C (1013 hPa)
Melting Pt:550 °C
Density:4,12 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-713-4
UN:1549
ADR:6.1,III
:GHS07
:GHS09
Related Information
Frequently Bought Together