

Aluminium sulphide 99+% (metals basis), pieces
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1302-81-4
:
Formula:Al₂S₃
MDL Number:MFCD00014162
:150.16 g/mol
Merck Index:14,367
Boiling Pt:1500 °C (1013 hPa)
Melting Pt:1100 °C
Density:2,02
Storage Temperature:Ambient
EINECS:215-109-0
UN:3134
ADR:4.3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together