Boron carbide ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 41049.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
CAS Number:12069-32-8
:
Formula:CB₄
MDL Number:MFCD00011520
:55.25 g/mol
Merck Index:12,01374
Boiling Pt:∼3500 °C (1013 hPa)
Melting Pt:2350 °C
Density:2,52 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:235-111-5