Order Entry
Northern Ireland
ContactUsLinkComponent
Strontium titanate substrate 5x5x1 mm, polished both sides 110 orientation
Strontium titanate substrate 5x5x1 mm, polished both sides 110 orientation
Catalog # 42671.KT
CAS Number:  
Strontium titanate substrate 5x5x1 mm, polished both sides 110 orientation
Catalog # 42671.KT
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :183.5 g/mol
Melting Pt:2080 °C
Density:5,11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:235-044-1
CAS Number:12060-59-2
Formula:O₃SrTi
MDL Number:MFCD00049554