

1-(2,4-Difluorophenyl)piperazine ≥99%
Catalog # L19393.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:115761-79-0
:
Formula:C₁₀H₁₂F₂N₂
MDL Number:MFCD00082588
:198.22 g/mol
Boiling Pt:108…111 °C (0,2 mmHg)
Melting Pt:74…76 °C
Storage Temperature:Ambient
UN:2923
ADR:8,II
:GHS06
:GHS05