Order Entry
Northern Ireland
ContactUsLinkComponent
1-(2-Methoxyethyl)piperazine
1-(2-Methoxyethyl)piperazine
Catalog # APOSOR6898-5G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Methoxyethyl)piperazine
Catalog # APOSOR6898-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :144.22 g/mol
UN:3267
ADR:8,III
CAS Number:13484-40-7
Formula:C₇H₁₆N₂O
MDL Number:MFCD00191214