Order Entry
Northern Ireland
ContactUsLinkComponent
1-(2-Methoxyethyl)piperazine 98%
Corrosive
1-(2-Methoxyethyl)piperazine 98%
Catalog # ACRO367110010
CAS Number:  
undefined
Corrosive
1-(2-Methoxyethyl)piperazine 98%
Catalog # ACRO367110010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4720 to 1.4750 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :144.22 g/mol
Flash Pt:87 °C
UN:2735
ADR:8,III
 :GHS05
CAS Number:13484-40-7
Formula:C₇H₁₆N₂O
MDL Number:MFCD00191214