3-Cyclohexyl-1-propanol 99%
Catalog # ACRO255880050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Clear colorless Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4650 to 1.4670 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
About this item
:
:142.24 g/mol
Boiling Pt:223 °C (1013 hPa)
Melting Pt:–60 °C
Density:0,91 g/cm³
Flash Pt:116 °C
EINECS:214-401-5
CAS Number:1124-63-6
Formula:C₉H₁₈O
MDL Number:MFCD00001528