4-Methoxybiphenyl 97%, Sigma-Aldrich®
Catalog # 374776-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to off-white powder, crystals, crystalline powder and/or chunks
- Infrared spectrum:Conforms to structure
- Gas liquid:96.5% (minimum)
Specification Test Results
About this item
Synonyms: Biphenyl-4-yl methyl ether, 4-Phenylanisole
:
:184.24 g/mol
Melting Pt:86...90 °C (lit.)
Storage Temperature:Room Temperature
EINECS:210-339-8
CAS Number:613-37-6
Formula:C₁₃H₁₂O
MDL Number:MFCD00014897