Order Entry
Northern Ireland
ContactUsLinkComponent
Oxybenzone
Oxybenzone
Catalog # MOLEM82203189
Supplier:  Molekula
CAS Number:  
Oxybenzone
Catalog # MOLEM82203189
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :228.25 g/mol
Merck Index:12,07088
Boiling Pt:370 °C (1013 hPa)
Melting Pt:65 °C
Density:1,3397 g/cm³ (25 °C)
EINECS:205-031-5
CAS Number:131-57-7
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00008387