Supelco® PESTANAL® Organic Reference Standard, 4,4′-DDD
35486-250MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:72-54-8
- Description:4,4′-DDD
- Storage conditions:2...30 °C
- Hazard class description:H301 - H312 - H351 - H410
- Shipping conditions:STD
- MDL No.:MFCD00000851
- M-Clarity quality level:MQ100
- Synonym(s):1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane; 4,4′-DDD; TDE
- Formula:(ClC₆H₄)₂CHCHCl₂
- Molecular weight:320,04
- GHS signal: GHS08, GHS06, GHS09
: P201 - P273 - P280 - P301 + P310 + P330 - P302 + P352 + P312