

2,4'-DDE, extra pure
Catalog # 123842Q
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:318.03 g/mol
Boiling Pt:64…65 °C (1013 hPa)
Melting Pt:–98 °C
Flash Pt:11 °C
Storage Temperature:Refrigerator
EINECS:222-318-0
UN:2930
ADR:6.1 (4.1),II
:GHS02
:GHS06
CAS Number:3424-82-6
Formula:C₁₄H₈Cl₄