
5-Aminoisophthalic acid for synthesis, Sigma-Aldrich®
8.01246.0050
: Merck
:
- Pack type:Plastic bottle
- Pk:50 g
- Seq:3368
- Solubility:(20 °C) insoluble
- Melting Point:150 - 156 °C
- Molar Mass:181.15 g/mol
:
:181.15 g/mol
:515 °C (1013 hPa)
:150…156 °C
:M-Clarity™ quality level = MQ200
:202-748-5
:GHS07
:99-31-0
:C₈H₇NO₄
:MFCD00002522