Order Entry
Northern Ireland
ContactUsLinkComponent
2-Chloro-4-methoxyphenol 97%
Irritant
2-Chloro-4-methoxyphenol 97%
Catalog # ACRO294100250
CAS Number:  
undefined
Irritant
2-Chloro-4-methoxyphenol 97%
Catalog # ACRO294100250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Low melting crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    38 to 43 °C
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :158.58 g/mol
Boiling Pt:242 °C (1013 hPa)
Melting Pt:38…43 °C
Flash Pt:85 °C
Storage Temperature:Ambient
EINECS:242-007-3
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:18113-03-6
Formula:C₇H₇ClO₂
MDL Number:MFCD00070773