Order Entry
Northern Ireland
ContactUsLinkComponent
1-Naphthyl butyrate solution, Sigma-Aldrich®
Flammable
Health
Irritant
1-Naphthyl butyrate solution, Sigma-Aldrich®
Catalog # 1801-50ML
CAS Number:  
Flammable
Health
Irritant
1-Naphthyl butyrate solution, Sigma-Aldrich®
Catalog # 1801-50ML
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
ADR:3,III
 :214.26 g/mol
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Freezer
 :1
CAS Number:3121-70-8
Formula:C₁₄H₁₄O₂
MDL Number:MFCD00003923