Order Entry
Northern Ireland
ContactUsLinkComponent
Diatrizoic acid, Sigma-Aldrich®
Diatrizoic acid, Sigma-Aldrich®
Catalog # D9268-50G
CAS Number:  
undefined
Diatrizoic acid, Sigma-Aldrich®
Catalog # D9268-50G
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :613.92 g/mol
Storage Temperature:Ambient
CAS Number:117-96-4
Formula:C₁₁H₉I₃N₂O₄
MDL Number:MFCD00069960