γ-(N-Morpholino)propanesulphonic acid (MOPS), Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:1132-61-2
:
Formula:C₇H₁₅NO₄S
MDL Number:MFCD00006183
:209.27 g/mol
Merck Index:13,06288
Melting Pt:> 250 °C
:M-Clarity™ quality level = MQ100
Flash Pt:110 °C (closed cup)
Storage Temperature:Ambient
EINECS:214-478-5
Frequently Bought Together