β-(N-Morpholino)ethanesulphonic acid (MES)
Catalog # PROOMM0334.03
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:195.24 g/mol
Merck Index:13,05929
Melting Pt:316 °C
Storage Temperature:Ambient
EINECS:224-632-3
CAS Number:4432-31-9
Formula:C₆H₁₃NO₄S
MDL Number:MFCD00006181