α-Linolenic acid methyl ester 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:301-00-8
:
Formula:C₁₉H₃₂O₂
MDL Number:MFCD00135851
:292.46 g/mol
Boiling Pt:182 °C (3 mmHg)
Melting Pt:–57 °C
Density:0,895 g/cm³
Flash Pt:>110 °C
Storage Temperature:Freezer
EINECS:206-102-3
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4700 to 1.4720 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together