Toluene-α-sulphonyl fluoride
Catalog # 786-055
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:174.2 g/mol
Boiling Pt:80…85 °C (0,3 mmHg)
Melting Pt:91…92 °C
ADR:8,III
CAS Number:329-98-6
Formula:C₇H₇FO₂S
MDL Number:MFCD00007424