
1,1,3,3-Tetramethoxypropane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:102-52-3
:
Formula:C₇H₁₆O₄
MDL Number:MFCD00008488
:164.2 g/mol
Boiling Pt:183 °C (1013 hPa)
Density:0,99 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-037-2
UN:1993
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Molar Mass:164.2 g/mol
- Boiling Point:183 °C
- Vapor Pressure:1.7 hPa (20 °C)
- Flash Point:66 °C
- Refractive Index:1.4067 (20 °C, 589 nm)
- Density:0.99 g/cm3 (20 °C)
- pH:7 (H2O)
- Viscosity kinematic:1.75 mm2/s (20 °C)
Specification Test Results
Frequently Bought Together