Lithium acetate dihydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6108-17-4
:
Formula:H₃CCOOLi·2H₂O
MDL Number:MFCD00066949
:102.02 g/mol
Merck Index:14,5521
Melting Pt:54…57 °C
Density:1,300
Storage Temperature:Ambient
EINECS:208-914-3
Related Information
Frequently Bought Together