


2-Methoxyethyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-49-6
:
Formula:C₅H₁₀O₃
MDL Number:MFCD00008719
:118.13 g/mol
Merck Index:12,06119
Boiling Pt:145 °C (1013 hPa)
Melting Pt:–65 °C
Density:1,01 g/cm³ (20 °C)
EINECS:203-772-9
UN:1189
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:49 °C
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-65 °C
- Molar Mass:118.13 g/mol
- Boiling Point:144 - 145 °C (1013 hPa)
- Vapor Pressure:4.7 hPa (20 °C)
- Flash Point:46 °C
- Refractive Index:1.4019 (20 °C, 589 nm)
- Explosion Limit:1.7 - 8.8 %(V)
- Density:1.01 g/cm3 (20 °C)
- Ignition Point:380 °C DIN 51794
- Evaporation Number:30
Specification Test Results
Frequently Bought Together