Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Methyl tridecanoate, Supelco®

Methyl tridecanoate, Supelco®

Glass bottle, 5 mL
Catalog # 91558-5ML
Supplier:   Merck
Methyl tridecanoate, Supelco®
Catalog # 91558-5ML
Supplier:  Merck
CAS Number:  1731-88-0
Methyl tridecanoate, Supelco®
Methyl tridecanoate, Supelco®

Glass bottle, 5 mL

CAS Number:  1731-88-0
Price
£218.00
Log in for availability
1 * 5 mL
Product Details & Documents
Synonyms:  Tridecylic acid methyl ester, Tridecanoic acid methyl ester
  • CAS Number:1731-88-0
  • Formula:C₁₄H₂₈O₂
  • MDL Number:MFCD00008977
  • Molecular Weight:228.38 g/mol
  • Boiling Pt:290 °C (1013 hPa)
  • Melting Pt:5,5 °C
  • Density:0,864 g/cm³ (25 °C)
  •  :M-Clarity™ quality level = MQ100
  • EINECS:217-054-8
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.