Ethyl tridecanoate for synthesis, Sigma-Aldrich®
Catalog # 8.08634.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 ml
- Seq:2275
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Molar Mass:242.4 g/mol
- Density:0.86 g/cm3 (20 °C)
- Synonyms:Tridecanoic acid ethyl ester
- Hill Formula:C15H30O2
- Chemical Formula:CH3(CH2)11COOC2H5
Specification Test Results
About this item
:
:242.4 g/mol
Density:0,86 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:248-929-2
CAS Number:28267-29-0
Formula:C₁₅H₃₀O₂
MDL Number:MFCD00008978