Bismuth(III) carbonate basic
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 Formula Weight: 509.97 Merck Reference: 14,1282 MDL No.: MFCD00010877
Synonyms: Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
- CAS Number:5892-10-4
- Formula:(BiO)₂CO₃
- MDL Number:MFCD00010877
- Molecular Weight:509.97 g/mol
- Merck Index:14,1282
- Density:6,86
- Storage Temperature:Ambient
- EINECS:227-567-9
3 Options Available Below
Product Details & Documents
CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 Formula Weight: 509.97 Merck Reference: 14,1282 MDL No.: MFCD00010877
Synonyms: Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
- CAS Number:5892-10-4
- Formula:(BiO)₂CO₃
- MDL Number:MFCD00010877
- Molecular Weight:509.97 g/mol
- Merck Index:14,1282
- Density:6,86
- Storage Temperature:Ambient
- EINECS:227-567-9
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



