Bismuth(III) carbonate basic ≥98.5%
About this item
Purity: 98.5% min CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 (approximate) Formula Weight: 509.97 Physical Form: Powder Density: 6.86 Solubility: Practically insoluble in water, alcohol. Soluble in nitric acid, hydrochloric acid, concentrated acetic acid Merck Reference: 14,1282 MDL No.: MFCD00010877
Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes
- CAS Number:5892-10-4
- Formula:(BiO)₂CO₃
- MDL Number:MFCD00010877
- Molecular Weight:509.97 g/mol
- Merck Index:14,1282
- Density:6,86
- Storage Temperature:Ambient
- EINECS:227-567-9
2 Options Available Below
Product Details & Documents
Purity: 98.5% min CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 (approximate) Formula Weight: 509.97 Physical Form: Powder Density: 6.86 Solubility: Practically insoluble in water, alcohol. Soluble in nitric acid, hydrochloric acid, concentrated acetic acid Merck Reference: 14,1282 MDL No.: MFCD00010877
Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes
- CAS Number:5892-10-4
- Formula:(BiO)₂CO₃
- MDL Number:MFCD00010877
- Molecular Weight:509.97 g/mol
- Merck Index:14,1282
- Density:6,86
- Storage Temperature:Ambient
- EINECS:227-567-9
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



