


(S)-(-)-1-Phenylethyl isocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14649-03-7
:
Formula:C₉H₉NO
MDL Number:MFCD00002036
:147.18 g/mol
Boiling Pt:55…56 °C (2,5 mmHg)
Density:1,045
Flash Pt:65 °C (149 °F)
EINECS:238-698-6
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together