
(S)-(-)-1-Phenylethyl isocyanate 98%
:14649-03-7
:
:C₉H₉NO
:MFCD00002036
:147.18 g/mol
:55…56 °C (2,5 mmHg)
:1,05 g/cm³
:65 °C
:Refrigerator
:238-698-6
:2206
:6.1,II
:GHS06
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5130 to 1.5150 (20°C, 589 nm)
- GC:≥97.5 %
- Specific optical rotation:-9° to -11° (20°C, 589 nm) (neat)
Frequently Bought Together