Cobalt(II) bromide hydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:85017-77-2
:
Formula:Br₂Co·H₂O
MDL Number:MFCD00150208
:236.76 g/mol
Density:2,46 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-166-7
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Violet Crystals
- Titration Argentometric:17.0 to 20.3 % (Co)
- Water:24.7 to 36.5 %
Specification Test Results
Frequently Bought Together