

Zirconyl nitrate hydrate 99.5% contains up to 4% Hf
:14985-18-3
:
:N₂O₇Zr·H₂O
:MFCD00149896
:249.25 g/mol
:2,04 g/cm³ (20 °C)
:237-529-3
:3085
:5.1,III
:GHS05
:GHS03
- Appearance:White Powder and/or chunks
- Degree of hydration:6 to 7
- Gravimetry:≥99.4 % (calculated on the actual degree of hydration)
- Hafnium (Hf):≤4 %
Frequently Bought Together