
Zirconyl chloride octahydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13520-92-8
:
Formula:Cl₂OZr·8H₂O
MDL Number:MFCD00149898
:322.25 g/mol
Melting Pt:> 150 °C
Density:1,91 g/cm³
Storage Temperature:Ambient
EINECS:231-717-9
UN:3260
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Titration Argentometric:97.5 to 102.5 %
- Water:42 to 50 %
- Solubility:(10% in water) Clear to slightly hazy solution
Specification Test Results
Frequently Bought Together