
Cerium(III) sulphate octahydrate 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10450-59-6
:
Formula:Ce₂O₁₂S₃·8H₂O
MDL Number:MFCD00149641
:712.54
EINECS:236-644-6
:GHS07
Specification Test Results
- Appearance:White to off-white Crystals and/or chunks
- Assay:38.0 to 40.5 % (Ce)
- Total trace metal impurities:≤0.01 %
Specification Test Results
Frequently Bought Together