

Lanthanum(III) chloride heptahydrate ≥99%, Puriss. p.a., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:10025-84-0
:
Formula:LaCl₃·7H₂O
MDL Number:MFCD00149756
Boiling Pt:? 860 °C (1013 hPa)
Melting Pt:91 °C (dec.)
Storage Temperature:Ambient
EINECS:233-237-5
UN:3260
ADR:8,III
:371.37 g/mol
:GHS07
:GHS05
Specification Test Results
- Assay:Min. 99 %
- Calcium (Ca):Max. 0.0005 %
- Iron (Fe):Max. 0.0005 %
- Potassium (K):Max. 0.001 %
- Magnesium (Mg):Max. 0.0005 %
- Sodium (Na):Max. 0.001 %
- Heavy metals (as Pb):Max. 0.0005 %
- Sulfate (SO₄):Max. 0.005 %
Specification Test Results
Frequently Bought Together