Gallium trinitrate hydrate ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69365-72-6
:
Formula:GaN₃O₉·H₂O
MDL Number:MFCD00149723
:273.75 g/mol
Merck Index:14,4346
Melting Pt:110 °C (dec.)
Storage Temperature:Ambient
EINECS:236-815-5
UN:1477
ADR:5.1,III
:GHS03
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together