Lithium sulphate monohydrate ≥99.0% (dried basis), white powder ACS
:10102-25-7
:
:Li₂SO₄·H₂O
:MFCD00149766
:2.05 g/cm³ (20 °C)
:127.96 g/mol
:120
:233-820-4
:GHS07
:Ambient
- Environmentally Preferable:
- Assay:≥99,0%
- Heavy Metals:≤0,001% (as Pb)
- Insoluble Matter:≤0,01%
- Loss on Drying:13,0 - 15,0% (150 °C)
- Chloride:≤0,002%
- Nitrate:≤0,001%
- Iron:<0,001%
- Potassium:≤0,05%
- Sodium:≤0,05%
Frequently Bought Together