

Disodium metasilicate pentahydrate, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10213-79-3
:
Formula:Na₂SiO₃·5H₂O
MDL Number:MFCD00149176
:212.14 g/mol
Merck Index:14,8642
Melting Pt:72 °C
Density:1,75
Storage Temperature:Ambient
EINECS:229-912-9
UN:3253
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together